Assay Details
| Gene Symbol: LOC4337592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783235.1 | XP_015638721.1 | 6 - 7 | 852 | 64 | |
| GenBank mRNA | AK067749.1 | - | 7 - 8 | 641 | 64 | |
| AK104935.1 | - | 3 - 4 | 267 | 64 | ||
More Information
Set Membership: |
|

LOC4337592
Genomic Map