Assay Details
| Gene Symbol: LOC4337640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783331.2 | XP_015638817.1 | 1 - 2 | 250 | 77 | |
| GenBank mRNA | AK110829.1 | - | 1 - 2 | 253 | 77 | |
More Information
Set Membership: |
|

LOC4337640
Genomic Map