Assay Details
| Gene Symbol: LOC4337658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782987.2 | XP_015638473.2 | 13 - 14 | 3531 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4337658| Gene Symbol: LOC4337658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782987.2 | XP_015638473.2 | 13 - 14 | 3531 | 78 | |
Set Membership: |
|