Assay Details
| Gene Symbol: LOC4337721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784054.2 | XP_015639540.1 | 10 - 11 | 1216 | 84 | |
| GenBank mRNA | AK063482.1 | - | 10 - 11 | 1216 | 84 | |
| AK067302.1 | - | 10 - 11 | 1217 | 84 | ||
More Information
Set Membership: |
|

LOC4337721
Genomic Map