Assay Details
| Gene Symbol: LOC4337721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784054.2 | XP_015639540.1 | 2 - 3 | 469 | 65 | |
| GenBank mRNA | AK063482.1 | - | 2 - 3 | 469 | 65 | |
| AK067302.1 | - | 2 - 3 | 470 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784054.2 transcripts.
Set Membership: |
|

LOC4337721
Genomic Map