Assay Details
| Gene Symbol: LOC4337721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784054.2 | XP_015639540.1 | 8 - 9 | 1036 | 113 | |
| GenBank mRNA | AK063482.1 | - | 8 - 9 | 1036 | 113 | |
| AK067302.1 | - | 8 - 9 | 1037 | 113 | ||
More Information
Set Membership: |
|

LOC4337721
Genomic Map