Assay Details
| Gene Symbol: LOC4337826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783896.2 | XP_015639382.1 | 2 - 3 | 430 | 132 | |
| XM_015783897.2 | XP_015639383.1 | 2 - 3 | 333 | 132 | ||
| XM_015783898.2 | XP_015639384.1 | 2 - 3 | 308 | 132 | ||
| GenBank mRNA | AK110983.1 | - | 2 - 3 | 357 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015783896.2, XM_015783897.2, XM_015783898.2 transcripts.
Set Membership: |
|

LOC4337826
Genomic Map