Assay Details
| Gene Symbol: LOC4337826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783896.2 | XP_015639382.1 | 3 - 4 | 680 | 82 | |
| XM_015783897.2 | XP_015639383.1 | 3 - 4 | 583 | 82 | ||
| XM_015783898.2 | XP_015639384.1 | 3 - 4 | 558 | 82 | ||
| GenBank mRNA | AK110983.1 | - | 3 - 4 | 607 | 82 | |
More Information
Set Membership: |
|

LOC4337826
Genomic Map