Assay Details
| Gene Symbol: LOC4337861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784371.2 | XP_015639857.1 | 3 - 4 | 714 | 77 | |
| GenBank mRNA | AK102557.1 | - | 3 - 4 | 668 | 77 | |
More Information
Set Membership: |
|

LOC4337861
Genomic Map