Assay Details
Gene Symbol: LOC4350997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK288106.1 | - | 2659 | 131 | ||
AK318564.1 | - | 2659 | 131 |
Gene Symbol: LOC4337867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784435.1 | XP_015639921.1 | 2 - 3 | 412 | 131 | |
GenBank mRNA | AK065771.1 | - | 2 - 3 | 391 | 131 | |
AK103075.1 | - | 2 - 3 | 391 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
