Assay Details
| Gene Symbol: LOC4338161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785017.2 | XP_015640503.1 | 3 - 4 | 513 | 76 | |
| GenBank mRNA | AK067996.1 | - | 2 - 3 | 259 | 76 | |
| HQ013294.1 | - | 2 - 3 | 270 | 76 | ||
More Information
Set Membership: |
|

LOC4338161
Genomic Map