Assay Details
| Gene Symbol: LOC4338287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783328.2 | XP_015638814.1 | 1 - 2 | 878 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4338287| Gene Symbol: LOC4338287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783328.2 | XP_015638814.1 | 1 - 2 | 878 | 94 | |
Set Membership: |
|