Assay Details
| Gene Symbol: LOC4338463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784827.2 | XP_015640313.1 | 8 - 9 | 1130 | 102 | |
| GenBank mRNA | AK243668.1 | - | 8 - 9 | 1131 | 102 | |
More Information
Set Membership: |
|

LOC4338463
Genomic Map