Assay Details
| Gene Symbol: LOC4338503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784088.2 | XP_015639574.2 | 1 | 684 | 89 | |
| GenBank mRNA | AK288128.1 | - | 1 | 79 | 89 | |
More Information
Set Membership: |
|

LOC4338503
Genomic Map