Assay Details
| Gene Symbol: LOC4338911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784687.2 | XP_015640173.1 | 2 - 3 | 493 | 66 | |
| GenBank mRNA | AK059951.1 | - | 2 - 3 | 343 | 66 | |
More Information
Set Membership: |
|

LOC4338911
Genomic Map