Assay Details
| Gene Symbol: LOC4338940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784881.2 | XP_015640367.1 | 13 - 14 | 5212 | 79 | |
| XM_015784882.2 | XP_015640368.1 | 13 - 14 | 5209 | 79 | ||
| XM_015784883.2 | XP_015640369.1 | 13 - 14 | 5218 | 79 | ||
| GenBank mRNA | AK103871.1 | - | 7 - 8 | 833 | 79 | |
| FJ536320.1 | - | 12 - 13 | 4849 | 79 | ||
More Information
Set Membership: |
|

LOC4338940
Genomic Map