Assay Details
| Gene Symbol: LOC4338942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782110.2 | XP_015637596.1 | 2 - 3 | 328 | 82 | |
| XM_015782111.2 | XP_015637597.1 | 2 - 3 | 338 | 82 | ||
| GenBank mRNA | AK066998.1 | - | 2 - 3 | 302 | 82 | |
More Information
Set Membership: |
|

LOC4338942
Genomic Map