Assay Details
| Gene Symbol: LOC4338942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782110.2 | XP_015637596.1 | 3 - 4 | 515 | 80 | |
| XM_015782111.2 | XP_015637597.1 | 3 - 4 | 525 | 80 | ||
| GenBank mRNA | AK066998.1 | - | 3 - 4 | 489 | 80 | |
More Information
Set Membership: |
|

LOC4338942
Genomic Map