Assay Details
| Gene Symbol: LOC4338971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781919.2 | XP_015637405.1 | 7 - 8 | 677 | 83 | |
| XM_015781920.2 | XP_015637406.1 | 7 - 8 | 849 | 83 | ||
| GenBank mRNA | AK102175.1 | - | 5 - 6 | 1973 | 83 | |
More Information
Set Membership: |
|

LOC4338971
Genomic Map