Assay Details
| Gene Symbol: LOC4339151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784927.2 | XP_015640413.1 | 2 - 3 | 326 | 100 | |
| XM_015784928.2 | XP_015640414.1 | 1 - 2 | 269 | 100 | ||
| XM_015784929.2 | XP_015640415.1 | 2 - 3 | 324 | 100 | ||
| XM_015784930.2 | XP_015640416.1 | 2 - 3 | 326 | 100 | ||
| XM_015784931.2 | XP_015640417.1 | 2 - 3 | 326 | 100 | ||
| GenBank mRNA | AK241362.1 | - | 1 - 2 | 269 | 100 | |
More Information
Set Membership: |
|

LOC4339151
Genomic Map