Assay Details
| Gene Symbol: LOC4339151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784927.2 | XP_015640413.1 | 5 - 6 | 550 | 58 | |
| XM_015784928.2 | XP_015640414.1 | 4 - 5 | 493 | 58 | ||
| XM_015784929.2 | XP_015640415.1 | 5 - 6 | 548 | 58 | ||
| XM_015784930.2 | XP_015640416.1 | 5 - 6 | 535 | 58 | ||
| XM_015784931.2 | XP_015640417.1 | 5 - 6 | 550 | 58 | ||
| GenBank mRNA | AK241362.1 | - | 4 - 5 | 493 | 58 | |
More Information
Set Membership: |
|

LOC4339151
Genomic Map