Assay Details
| Gene Symbol: LOC4339173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783664.2 | XP_015639150.1 | 1 - 2 | 564 | 118 | |
| GenBank mRNA | AB110221.1 | - | 1 - 2 | 412 | 118 | |
| AK068637.1 | - | 1 - 2 | 565 | 118 | ||
| AK071229.1 | - | 1 - 2 | 417 | 118 | ||
More Information
Set Membership: |
|

LOC4339173
Genomic Map