Assay Details
| Gene Symbol: LOC4339173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783664.2 | XP_015639150.1 | 2 - 3 | 752 | 140 | |
| XM_015783665.2 | XP_015639151.1 | 2 - 3 | 647 | 140 | ||
| GenBank mRNA | AB110221.1 | - | 2 - 3 | 600 | 140 | |
| AK068637.1 | - | 2 - 3 | 753 | 140 | ||
| AK071229.1 | - | 2 - 3 | 605 | 140 | ||
More Information
Set Membership: |
|

LOC4339173
Genomic Map