Assay Details
| Gene Symbol: LOC4339221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783656.2 | XP_015639142.1 | 1 | 618 | 71 | |
| GenBank mRNA | AK242943.1 | - | 1 | 608 | 71 | |
| EF122478.1 | - | 1 | 588 | 71 | ||
| GQ848069.1 | - | 1 | 578 | 71 | ||
| X63990.1 | - | 1 | 562 | 71 | ||
| Gene Symbol: LOC107279298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287978.1 | - | 606 | 71 | ||
| Gene Symbol: LOC4344664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK242017.1 | - | 608 | 71 | ||
| Gene Symbol: LOC4344973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287940.1 | - | 608 | 71 | ||
More Information
Set Membership: |
|

LOC4339221
Genomic Map