Assay Details
| Gene Symbol: LOC4339599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784323.2 | XP_015639809.1 | 1 - 2 | 1603 | 74 | |
| GenBank mRNA | AK067862.1 | - | 1 - 2 | 1177 | 74 | |
| AK122091.1 | - | 1 - 2 | 1609 | 74 | ||
More Information
Set Membership: |
|

LOC4339599
Genomic Map