Assay Details
| Gene Symbol: LOC4339654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782006.2 | XP_015637492.1 | 4 - 5 | 1017 | 113 | |
| GenBank mRNA | AK100553.1 | - | 3 - 4 | 818 | 113 | |
More Information
Set Membership: |
|

LOC4339654
Genomic Map