Assay Details
| Gene Symbol: LOC4339683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783980.2 | XP_015639466.1 | 1 - 2 | 145 | 79 | |
| GenBank mRNA | AB015287.1 | - | 1 - 2 | 83 | 79 | |
| AB376668.1 | - | 1 - 2 | 62 | 79 | ||
| AK061714.1 | - | 1 - 2 | 101 | 79 | ||
| AK098889.1 | - | 1 - 2 | 97 | 79 | ||
| AK121699.1 | - | 1 - 2 | 102 | 79 | ||
| AK288455.1 | - | 2772 | 79 | |||
| BT129306.1 | - | 1 - 2 | 95 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015783980.2 transcripts.
Set Membership: |
|

LOC4339683
Genomic Map