Assay Details
| Gene Symbol: LOC4339683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783980.2 | XP_015639466.1 | 2 - 3 | 204 | 82 | |
| GenBank mRNA | AB015287.1 | - | 2 - 3 | 142 | 82 | |
| AB015972.1 | - | 2 - 3 | 115 | 82 | ||
| AB376668.1 | - | 2 - 3 | 121 | 82 | ||
| AK061714.1 | - | 2 - 3 | 160 | 82 | ||
| AK098889.1 | - | 2 - 3 | 156 | 82 | ||
| AK121699.1 | - | 2 - 3 | 161 | 82 | ||
| AK288455.1 | - | 2831 | 82 | |||
| BT129306.1 | - | 2 - 3 | 154 | 82 | ||
More Information
Set Membership: |
|

LOC4339683
Genomic Map