Assay Details
| Gene Symbol: LOC4339683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783980.2 | XP_015639466.1 | 4 - 5 | 387 | 110 | |
| GenBank mRNA | AB015287.1 | - | 4 - 5 | 325 | 110 | |
| AB376668.1 | - | 4 - 5 | 304 | 110 | ||
| AK061714.1 | - | 4 - 5 | 343 | 110 | ||
| AK098889.1 | - | 4 - 5 | 339 | 110 | ||
| AK288455.1 | - | 3014 | 110 | |||
| BT129306.1 | - | 4 - 5 | 337 | 110 | ||
| BT130326.1 | - | 1 - 2 | 42 | 110 | ||
More Information
Set Membership: |
|

LOC4339683
Genomic Map