Assay Details
| Gene Symbol: LOC4339683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783980.2 | XP_015639466.1 | 6 - 7 | 654 | 75 | |
| GenBank mRNA | AB015287.1 | - | 6 - 7 | 592 | 75 | |
| AB015972.1 | - | 6 - 7 | 565 | 75 | ||
| AB376668.1 | - | 6 - 7 | 571 | 75 | ||
| AK061714.1 | - | 6 - 7 | 610 | 75 | ||
| AK098889.1 | - | 6 - 7 | 606 | 75 | ||
| AK121699.1 | - | 6 - 7 | 611 | 75 | ||
| AK288455.1 | - | 3281 | 75 | |||
| BT129306.1 | - | 6 - 7 | 604 | 75 | ||
| BT130326.1 | - | 3 - 4 | 309 | 75 | ||
| BT130804.1 | - | 1 - 2 | 78 | 75 | ||
| BT131738.1 | - | 2 - 3 | 282 | 75 | ||
More Information
Set Membership: |
|

LOC4339683
Genomic Map