Assay Details
| Gene Symbol: LOC4339683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783980.2 | XP_015639466.1 | 7 - 8 | 739 | 88 | |
| GenBank mRNA | AB015287.1 | - | 7 - 8 | 677 | 88 | |
| AB015972.1 | - | 7 - 8 | 650 | 88 | ||
| AB376668.1 | - | 7 - 8 | 656 | 88 | ||
| AK061714.1 | - | 7 - 8 | 695 | 88 | ||
| AK098889.1 | - | 7 - 8 | 691 | 88 | ||
| AK121699.1 | - | 7 - 8 | 696 | 88 | ||
| AK288455.1 | - | 3366 | 88 | |||
| BT129306.1 | - | 7 - 8 | 689 | 88 | ||
| BT130326.1 | - | 4 - 5 | 394 | 88 | ||
| BT130804.1 | - | 2 - 3 | 163 | 88 | ||
| BT131738.1 | - | 3 - 4 | 367 | 88 | ||
More Information
Set Membership: |
|

LOC4339683
Genomic Map