Assay Details
| Gene Symbol: LOC4339796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784199.1 | XP_015639685.1 | 2 - 3 | 2790 | 110 | |
| GenBank mRNA | AK073484.1 | - | 3 - 4 | 2475 | 110 | |
More Information
Set Membership: |
|

LOC4339796
Genomic Map