Assay Details
| Gene Symbol: LOC4339796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784199.1 | XP_015639685.1 | 4 - 5 | 2991 | 92 | |
| GenBank mRNA | AK073484.1 | - | 5 - 6 | 2676 | 92 | |
More Information
Set Membership: |
|

LOC4339796
Genomic Map