Assay Details
| Gene Symbol: LOC4339859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785378.2 | XP_015640864.1 | 1 - 2 | 232 | 107 | |
| GenBank mRNA | AK071985.1 | - | 1 - 2 | 82 | 107 | |
| AK103793.1 | - | 1 - 2 | 101 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015785378.2 transcripts.
Set Membership: |
|

LOC4339859
Genomic Map