Assay Details
| Gene Symbol: LOC4339870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786701.2 | XP_015642187.1 | 2 - 3 | 437 | 90 | |
| GenBank mRNA | AK069627.1 | - | 2 - 3 | 428 | 90 | |
More Information
Set Membership: |
|

LOC4339870
Genomic Map