Assay Details
| Gene Symbol: LOC4339893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787455.2 | XP_015642941.1 | 3 - 4 | 1042 | 132 | |
| XM_015787456.2 | XP_015642942.1 | 3 - 4 | 1042 | 132 | ||
| XM_015787457.2 | XP_015642943.1 | 3 - 4 | 1042 | 132 | ||
| XM_015787459.2 | XP_015642945.1 | 3 - 4 | 1043 | 132 | ||
| XM_015787460.2 | XP_015642946.1 | 3 - 4 | 1043 | 132 | ||
| XM_015787461.2 | XP_015642947.1 | 3 - 4 | 1043 | 132 | ||
| XM_015787462.2 | XP_015642948.1 | 3 - 4 | 1072 | 132 | ||
| GenBank mRNA | AK065637.1 | - | 2 - 3 | 564 | 132 | |
More Information
Set Membership: |
|

LOC4339893
Genomic Map