Assay Details
| Gene Symbol: LOC4340018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789000.2 | XP_015644486.1 | 9 - 10 | 2623 | 63 | |
| XM_015789001.2 | XP_015644487.1 | 10 - 11 | 1500 | 63 | ||
| XM_015789002.2 | XP_015644488.1 | 10 - 11 | 1494 | 63 | ||
| XM_015789003.2 | XP_015644489.1 | 10 - 11 | 1432 | 63 | ||
| XM_015789004.2 | XP_015644490.1 | 9 - 10 | 2617 | 63 | ||
| XM_015789005.2 | XP_015644491.1 | 10 - 11 | 1488 | 63 | ||
| XM_015789006.2 | XP_015644492.1 | 10 - 11 | 1424 | 63 | ||
| GenBank mRNA | AB066093.1 | - | 9 - 10 | 1325 | 63 | |
| AB066094.1 | - | 9 - 10 | 1325 | 63 | ||
| AF515481.1 | - | 9 - 10 | 1301 | 63 | ||
| AF515482.1 | - | 9 - 10 | 1301 | 63 | ||
| AF515483.1 | - | 9 - 10 | 1301 | 63 | ||
| AK070431.1 | - | 10 - 11 | 1472 | 63 | ||
| EU735072.1 | - | 9 - 10 | 1301 | 63 | ||
| KC332294.1 | - | 9 - 10 | 1301 | 63 | ||
| KT199369.1 | - | 10 - 11 | 1371 | 63 | ||
More Information
Set Membership: |
|

LOC4340018
Genomic Map