Assay Details
| Gene Symbol: LOC4340018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789004.2 | XP_015644490.1 | 7 - 8 | 2199 | 78 | |
| XM_015789005.2 | XP_015644491.1 | 8 - 9 | 1070 | 78 | ||
| XM_015789006.2 | XP_015644492.1 | 8 - 9 | 1006 | 78 | ||
| GenBank mRNA | AB066093.1 | - | 7 - 8 | 907 | 78 | |
| AB066094.1 | - | 7 - 8 | 907 | 78 | ||
| AF515481.1 | - | 7 - 8 | 883 | 78 | ||
| AF515482.1 | - | 7 - 8 | 883 | 78 | ||
| AF515483.1 | - | 7 - 8 | 883 | 78 | ||
| AK070431.1 | - | 8 - 9 | 1054 | 78 | ||
| EU735072.1 | - | 7 - 8 | 883 | 78 | ||
| KC332294.1 | - | 7 - 8 | 883 | 78 | ||
| KT199369.1 | - | 8 - 9 | 953 | 78 | ||
More Information
Set Membership: |
|

LOC4340018
Genomic Map