Assay Details
| Gene Symbol: LOC4340089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787008.1 | XP_015642494.1 | 2 - 3 | 461 | 96 | |
| GenBank mRNA | AK073269.1 | - | 2 - 3 | 428 | 96 | |
| AK103971.1 | - | 2 - 3 | 424 | 96 | ||
More Information
Set Membership: |
|

LOC4340089
Genomic Map