Assay Details
| Gene Symbol: LOC4340170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787707.2 | XP_015643193.1 | 4 - 5 | 991 | 122 | |
| GenBank mRNA | AB183398.1 | - | 4 - 5 | 932 | 122 | |
| AJ535841.1 | - | 4 - 5 | 884 | 122 | ||
| AK111691.1 | - | 4 - 5 | 922 | 122 | ||
| AK111942.1 | - | 4 - 5 | 923 | 122 | ||
More Information
Set Membership: |
|

LOC4340170
Genomic Map