Assay Details
| Gene Symbol: LOC4340258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785565.2 | XP_015641051.1 | 16 - 17 | 2622 | 78 | |
| GenBank mRNA | AK068858.1 | - | 16 - 17 | 2598 | 78 | |
| EU602344.1 | - | 16 - 17 | 2598 | 78 | ||
More Information
Set Membership: |
|

LOC4340258
Genomic Map