Assay Details
| Gene Symbol: LOC4340258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785565.2 | XP_015641051.1 | 2 - 3 | 572 | 72 | |
| GenBank mRNA | AK068858.1 | - | 2 - 3 | 548 | 72 | |
| EU602344.1 | - | 2 - 3 | 548 | 72 | ||
More Information
Set Membership: |
|

LOC4340258
Genomic Map