Assay Details
| Gene Symbol: LOC9268329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287768.1 | - | 1 | 53 | 61 | |
| Gene Symbol: LOC4340333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787814.2 | XP_015643300.1 | 1 | 1934 | 61 | |
| GenBank mRNA | AK109842.1 | - | 1 | 1346 | 61 | |
| KX430016.1 | - | 1 | 1291 | 61 | ||
More Information
Set Membership: |
|

LOC9268329
Genomic Map