Assay Details
| Gene Symbol: LOC112936043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785640.2 | XP_015641126.1 | 5 - 6 | 827 | 98 | |
| GenBank mRNA | AK071932.1 | - | 6 - 7 | 765 | 98 | |
More Information
Set Membership: |
|

LOC112936043
Genomic Map