Assay Details
| Gene Symbol: LOC4340810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787578.2 | XP_015643064.1 | 7 - 8 | 2943 | 127 | |
| GenBank mRNA | AK060808.1 | - | 3 - 4 | 453 | 127 | |
| AK065437.1 | - | 6 - 7 | 3084 | 127 | ||
More Information
Set Membership: |
|

LOC4340810
Genomic Map