Assay Details
| Gene Symbol: LOC4340810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787578.2 | XP_015643064.1 | 9 - 10 | 3378 | 69 | |
| GenBank mRNA | AK060808.1 | - | 5 - 6 | 888 | 69 | |
| AK065437.1 | - | 8 - 9 | 3519 | 69 | ||
More Information
Set Membership: |
|

LOC4340810
Genomic Map