Assay Details
| Gene Symbol: LOC4340928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785809.2 | XP_015641295.1 | 4 - 5 | 1234 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4340928| Gene Symbol: LOC4340928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785809.2 | XP_015641295.1 | 4 - 5 | 1234 | 128 | |
Set Membership: |
|