Assay Details
| Gene Symbol: LOC4341280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787528.2 | XP_015643014.1 | 15 - 16 | 2731 | 208 | |
| XM_015787529.2 | XP_015643015.1 | 14 - 15 | 2605 | 208 | ||
| GenBank mRNA | AK064200.1 | - | 2 - 3 | 329 | 115 | |
More Information
Set Membership: |
|

LOC4341280
Genomic Map