Assay Details
| Gene Symbol: LOC4341280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787528.2 | XP_015643014.1 | 17 - 18 | 3161 | 113 | |
| XM_015787529.2 | XP_015643015.1 | 16 - 17 | 3035 | 113 | ||
| GenBank mRNA | AK064200.1 | - | 5 - 6 | 666 | 113 | |
More Information
Set Membership: |
|

LOC4341280
Genomic Map