Assay Details
| Gene Symbol: LOC4341648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788830.2 | XP_015644316.1 | 9 - 10 | 1355 | 73 | |
| XM_015788831.2 | XP_015644317.1 | 9 - 10 | 1350 | 73 | ||
| GenBank mRNA | AK101292.1 | - | 9 - 10 | 1361 | 73 | |
| AK101919.1 | - | 9 - 10 | 1328 | 73 | ||
More Information
Set Membership: |
|

LOC4341648
Genomic Map